accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH3COCH2

CAS RN 2675-94-7

IUPAC Name: N,N-diethylprop-2-enamide
Synonyms: N,N-diethylprop-2-enamide
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: OVHHHVAVHBHXAK-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results