accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26759-46-6

IUPAC Name: methyl 2-amino-4,5-dimethoxybenzoate
Synonyms: methyl 2-amino-4,5-dimethoxybenzoate
Molecular Weight (g/mol): 211.22
Molecular Formula: C10H13NO4
InChi Key: QQFHCCQSCQBKBG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=C(OC)C=C1N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results