accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26819-07-8

IUPAC Name: N-ethyl-3-methylbenzamide
Synonyms: benzamide, N-ethyl-3-methyl-
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: FPINATACRXASTP-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=CC=CC(C)=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results