accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNHNHNN

CAS RN 2683-90-1

IUPAC Name: 2H,3H,7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one
Synonyms: 2H,3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one
Molecular Weight (g/mol): 137.1
Molecular Formula: C4H3N5O
InChi Key: OEEYCNOOAHGFHL-UHFFFAOYSA-N
SMILES: O=C1N=CN=C2NNN=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results