accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 26830-96-6

IUPAC Name: 2-amino-4-methylbenzonitrile
Synonyms: 2-amino-4-methylbenzonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: LGNVAEIITHYWCG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C#N)C(N)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results