accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cl(E)

CAS RN 2687-12-9

IUPAC Name: [(1E)-3-chloroprop-1-en-1-yl]benzene
Synonyms: [(1E)-3-chloroprop-1-en-1-yl]benzene
Molecular Weight (g/mol): 152.62
Molecular Formula: C9H9Cl
InChi Key: IWTYTFSSTWXZFU-QPJJXVBHSA-N
SMILES: ClC\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)