accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2O

CAS RN 2687-43-6

IUPAC Name: hydrogen O-benzylhydroxylamine chloride
Synonyms: hydrogen O-benzylhydroxylamine chloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: HYDZPXNVHXJHBG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NOCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)