accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3

CAS RN 2690-08-6

IUPAC Name: 1-(octylsulfanyl)octane
Synonyms: 1-(octylsulfanyl)octane
Molecular Weight (g/mol): 258.51
Molecular Formula: C16H34S
InChi Key: LOXRGHGHQYWXJK-UHFFFAOYSA-N
SMILES: CCCCCCCCSCCCCCCCC
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results