accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COBOH3CH3COH

CAS RN 269409-70-3

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 220.08
Molecular Formula: C12H17BO3
InChi Key: BICZJRAGTCRORZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)