accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 269409-99-6

IUPAC Name: ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 276.14
Molecular Formula: C15H21BO4
InChi Key: RFBZWPFBCXBBJS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results