accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH2

CAS RN 2695-47-8

IUPAC Name: 6-bromohex-1-ene
Synonyms: 6-bromohex-1-ene
Molecular Weight (g/mol): 163.06
Molecular Formula: C6H11Br
InChi Key: RIMXEJYJXDBLIE-UHFFFAOYSA-N
SMILES: BrCCCCC=C
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)