accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2700-30-3

H3CCH3NOCH3H3C

CAS RN 2700-30-3

IUPAC Name: N,N-bis(propan-2-yl)formamide
Synonyms: N,N-diisopropylformamide
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: UNBDDZDKBWPHAX-UHFFFAOYSA-N
SMILES: CC(C)N(C=O)C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)