accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27025-25-8

H3COOOOCH3NH2HCl

CAS RN 27025-25-8

IUPAC Name: 1,5-dimethyl 2-aminopentanedioate hydrochloride
Synonyms: 1,5-dimethyl 2-aminopentanedioate hydrochloride
Molecular Weight (g/mol): 211.64
Molecular Formula: C7H14ClNO4
InChi Key: MFUPLHQOVIUESQ-UHFFFAOYNA-N
SMILES: Cl.COC(=O)CCC(N)C(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)