accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27034-51-1

H3COOHNCl

CAS RN 27034-51-1

IUPAC Name: ethyl 6-chloro-1H-indole-2-carboxylate
Synonyms: ethyl 6-chloro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 223.66
Molecular Formula: C11H10ClNO2
InChi Key: FSMZLIBWSAMADK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=C(Cl)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results