accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27035-41-2

IUPAC Name: 5-methyl-1-benzothiophene-2-carbaldehyde
Synonyms: 5-methyl-1-benzothiophene-2-carbaldehyde
Molecular Weight (g/mol): 176.23
Molecular Formula: C10H8OS
InChi Key: SIHFZJLPKQUYAH-UHFFFAOYSA-N
SMILES: CC1=CC=C2SC(C=O)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results