CAS RN 27046-19-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Oxo Cyclophosphamide, TRC
CAS: 27046-19-1 Molecular Formula: C7 H12 Cl2 N O4 P Molecular Weight (g/mol): 276.05 IUPAC Name: 2-[bis(2-chloroethyl)amino]-2-oxo-1,3,2λ^{5}-dioxaphosphinan-5-one SMILES: ClCCN(CCCl)P1(=O)OCC(=O)CO1