accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27048-01-7

IUPAC Name: [3-(2-iodoacetamido)-2,2,5,5-tetramethylpyrrolidin-1-yl]oxidanyl
Synonyms: 3-(2-iodoacetamido)-2,2,5,5-tetramethylpyrrolidin-1-yloxidanyl
Molecular Weight (g/mol): 325.17
Molecular Formula: C10H18IN2O2
InChi Key: OQIGMSGDHDTSFA-UHFFFAOYNA-N
SMILES: CC1(C)CC(NC(=O)CI)C(C)(C)N1[O]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)