accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27058-83-9

IUPAC Name: hydrogen 4-chloro-1,3-benzothiazol-2-amine bromide
Synonyms: hydrogen 2-benzothiazolamine, 4-chloro- bromide
Molecular Weight (g/mol): 265.55
Molecular Formula: C7H6BrClN2S
InChi Key: DXXACNSRSDYMOR-UHFFFAOYSA-N
SMILES: [H+].[Br-].NC1=NC2=C(Cl)C=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results