accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2706-56-1

NH2N

CAS RN 2706-56-1

IUPAC Name: 2-(pyridin-2-yl)ethan-1-amine
Synonyms: 2-pyridylethylamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: XPQIPUZPSLAZDV-UHFFFAOYSA-N
SMILES: NCCC1=CC=CC=N1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)