accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2706-90-3

IUPAC Name: nonafluoropentanoic acid
Synonyms: nonafluoropentanoic acid
Molecular Weight (g/mol): 264.05
Molecular Formula: C5HF9O2
InChi Key: CXZGQIAOTKWCDB-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results