accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27072-45-3

IUPAC Name: 3',6'-dihydroxy-6-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: fluorescein isothiocyanate F6itc 6-fitc
Molecular Weight (g/mol): 389.38
Molecular Formula: C21H11NO5S
InChi Key: GTQFZXYECNSNNC-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=C2C=C(C=C3)N=C=S)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results