accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFIIF

CAS RN 2708-97-6

IUPAC Name: 1,2,3,4-tetrafluoro-5,6-diiodobenzene
Synonyms: 1,2,3,4-tetrafluoro-5,6-diiodobenzene
Molecular Weight (g/mol): 401.87
Molecular Formula: C6F4I2
InChi Key: JQBYIZAYQMMVTO-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(I)C(I)=C1F

 No Refinements