accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 271-73-8

IUPAC Name: 1H-pyrazolo[3,4-b]pyridine
Synonyms: 1H-pyrazolo[3,4-b]pyridine
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: GVLRTOYGRNLSDW-UHFFFAOYSA-N
SMILES: N1N=CC2=CC=CN=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results