accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFOOIOOFFF

CAS RN 2712-78-9

IUPAC Name: phenyl[(2,2,2-trifluoroacetyl)oxy]-λ³-iodanyl 2,2,2-trifluoroacetate
Synonyms: (bis(trifluoroacetoxy)iodo)benzene
Molecular Weight (g/mol): 430.04
Molecular Formula: C10H5F6IO4
InChi Key: PEZNEXFPRSOYPL-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)