accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOOOIOH

CAS RN 27126-76-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 392.21
Molecular Formula: C13H13IO4S
InChi Key: LRIUKPUCKCECPT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)O[I](O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)