accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27139-97-5

CH3ClBr

CAS RN 27139-97-5

IUPAC Name: 2-bromo-4-chloro-1-methylbenzene
Synonyms: 2-bromo-4-chloro-1-methylbenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: CSUUXPHPCXHYGY-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)