CAS RN 27144-18-9
IUPAC Name:
3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms:
3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol):
348.46
Molecular Formula:
C22H20O2S
InChi Key:
AECGEIVNZGQBJT-UHFFFAOYSA-N
SMILES:
OC(=O)CCSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
of
2
results
2
3-Tritylsulfanylpropionic Acid, TRC
CAS: 27144-18-9 Molecular Formula: C22H20O2S Molecular Weight (g/mol): 348.46 Synonym: 3-[(Triphenylmethyl)thio]propanoic Acid,3-(Triphenylmethylthio)propionic Acid,3-(Tritylthio)propanoic Acid,3-(Tritylthio)propionic Acid,3-[(Triphenylmethyl)mercapto]propionic Acid,NSC 96707,S-Trityl-3-mercaptopropionic Acid,β-(Tritylmercapto)propionic Acid IUPAC Name: 3-tritylsulfanylpropanoic acid SMILES: OC(=O)CCSC(c1ccccc1)(c2ccccc2)c3ccccc3