accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 2719-52-0

IUPAC Name: (pentan-2-yl)benzene
Synonyms: S-amylbenzene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: LTHAIAJHDPJXLG-UHFFFAOYNA-N
SMILES: CCCC(C)C1=CC=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results