accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNN

CAS RN 272-97-9

IUPAC Name: 3H-imidazo[4,5-c]pyridine
Synonyms: 3H-imidazo[4,5-c]pyridine
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: UBOOKRVGOBKDMM-UHFFFAOYSA-N
SMILES: N1C=NC2=CC=NC=C12
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)