accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH2NNHBr

CAS RN 27246-81-7

IUPAC Name: hydrogen (3-bromophenyl)hydrazine chloride
Synonyms: hydrogen (3-bromophenyl)hydrazine chloride
Molecular Weight (g/mol): 223.5
Molecular Formula: C6H8BrClN2
InChi Key: RPYIPFXHIKXRKS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results