accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2728-05-4

IUPAC Name: N,N-diethyl-4-methylbenzamide
Synonyms: N,N-diethyl-4-methylbenzamide
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: PUZORFQMRDHKBT-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results