accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClClF

CAS RN 2729-36-4

IUPAC Name: 4,5-dichloro-2-fluoroaniline
Synonyms: 4,5-dichloro-2-fluoroaniline
Molecular Weight (g/mol): 180
Molecular Formula: C6H4Cl2FN
InChi Key: FHJVHVKVTUTGTA-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results