accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27292-49-5

IUPAC Name: 3-(morpholin-4-yl)phenol
Synonyms: 3-(morpholin-4-yl)phenol
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: BMGSGGYIUOQZBZ-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)N1CCOCC1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)