accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H2N

CAS RN 27298-98-2

IUPAC Name: 1-(4-methylphenyl)ethan-1-amine
Synonyms: 1-(4-methylphenyl)ethanamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: UZDDXUMOXKDXNE-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C(C)C=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)