accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 273-21-2

IUPAC Name: 1H-imidazo[4,5-b]pyridine
Synonyms: 3H-imidazo[4,5-b]pyridine 1-deazapurine 7-azabenzimidazole azabenzimidazole
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: GAMYYCRTACQSBR-UHFFFAOYSA-N
SMILES: N1C=NC2=NC=CC=C12
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results