accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 273-53-0

IUPAC Name: 1,3-benzoxazole
Synonyms: benzoxazole
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: BCMCBBGGLRIHSE-UHFFFAOYSA-N
SMILES: O1C=NC2=CC=CC=C12
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)