accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 273221-94-6

CH3H3CCH3OON(R)(S)HOON

CAS RN 273221-94-6

IUPAC Name: (2S,4R)-1-[(tert-butoxy)carbonyl]-4-cyanopyrrolidine-2-carboxylic acid
Synonyms: (2S,4R)-1-(tert-butoxycarbonyl)-4-cyanopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 240.26
Molecular Formula: C11H16N2O4
InChi Key: MIVXQYMYEMUMKK-YUMQZZPRSA-N
SMILES: CC(C)(C)OC(=O)N1C[C@@H](C[C@H]1C(O)=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results