accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2736-40-5

IUPAC Name: 2-ethylbutanoyl chloride
Synonyms: butanoyl chloride, 2-ethyl-
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: SMUKODJVMQOSAB-UHFFFAOYSA-N
SMILES: CCC(CC)C(Cl)=O
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results