accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HBOO

CAS RN 274-07-7

IUPAC Name: 2H-1,3,2-benzodioxaborole
Synonyms: catecholborane
Molecular Weight (g/mol): 119.91
Molecular Formula: C6H5BO2
InChi Key: CENMEJUYOOMFFZ-UHFFFAOYSA-N
SMILES: B1OC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)