accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO(R)OO(R)OOOHO

CAS RN 2743-38-6

IUPAC Name: (2R,3R)-2,3-bis(benzoyloxy)butanedioic acid
Synonyms: (2R,3R)-2,3-bis(benzoyloxy)butanedioic acid
Molecular Weight (g/mol): 358.3
Molecular Formula: C18H14O8
InChi Key: YONLFQNRGZXBBF-ZIAGYGMSSA-N
SMILES: OC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results