accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONH2OCH3

CAS RN 27492-84-8

IUPAC Name: methyl 4-amino-2-methoxybenzoate
Synonyms: methyl 4-amino-2-methoxybenzoate
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: YUPQMVSYNJQULF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C=C1OC
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results