accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27503-81-7

IUPAC Name: 2-phenyl-1H-1,3-benzodiazole-6-sulfonic acid
Synonyms: phenylbenzimidazole sulfonic acid
Molecular Weight (g/mol): 274.29
Molecular Formula: C13H10N2O3S
InChi Key: UVCJGUGAGLDPAA-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results