accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrP

CAS RN 2751-90-8

IUPAC Name: tetraphenylphosphanium bromide
Synonyms: tetraphenylphosphonium bromide
Molecular Weight (g/mol): 419.3
Molecular Formula: C24H20BrP
InChi Key: BRKFQVAOMSWFDU-UHFFFAOYSA-M
SMILES: [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results