accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NONHNH2NH2NONHNH2NOHSOHOO

CAS RN 27511-79-1

IUPAC Name: bis(5-amino-1H-pyrazole-4-carboxamide); sulfuric acid
Synonyms: bis(3-amino-2H-pyrazole-4-carboxamide); sulfuric acid
Molecular Weight (g/mol): 350.31
Molecular Formula: C8H14N8O6S
InChi Key: UMPKASYMNORSRO-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC(=O)C1=C(N)NN=C1.NC(=O)C1=C(N)NN=C1