accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH3NCH3CH3Br

CAS RN 2758-06-7

IUPAC Name: (2-bromoethyl)trimethylazanium bromide
Synonyms: (2-bromoethyl)trimethylazanium bromide
Molecular Weight (g/mol): 246.97
Molecular Formula: C5H13Br2N
InChi Key: OINMNSFDYTYXEQ-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)CCBr
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results