accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiCH3H3COSOOFFF

CAS RN 27607-77-8

IUPAC Name: trimethylsilyl trifluoromethanesulfonate
Synonyms: trimethylsilyl triflate
Molecular Weight (g/mol): 222.25
Molecular Formula: C4H9F3O3SSi
InChi Key: FTVLMFQEYACZNP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OS(=O)(=O)C(F)(F)F
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)