accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SClSOOCl

CAS RN 2766-74-7

IUPAC Name: 5-chlorothiophene-2-sulfonyl chloride
Synonyms: 5-chlorothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 217.08
Molecular Formula: C4H2Cl2O2S2
InChi Key: SORSTNOXGOXWAO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(S1)S(Cl)(=O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results