accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNS

CAS RN 27685-94-5

IUPAC Name: 4-chlorothieno[3,2-c]pyridine
Synonyms: 4-chlorothieno[3,2-c]pyridine
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: VFPFMOXMHVQFNR-UHFFFAOYSA-N
SMILES: ClC1=C2C=CSC2=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results