accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 276861-63-3

H3COOOHFFF

CAS RN 276861-63-3

IUPAC Name: 3-methoxy-4-(trifluoromethyl)benzoic acid
Synonyms: 3-methoxy-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: HBMINIVKUUVKEB-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results