accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 276872-86-7

NOOO

CAS RN 276872-86-7

IUPAC Name: benzyl 3-formylpyrrolidine-1-carboxylate
Synonyms: benzyl 3-formylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 233.27
Molecular Formula: C13H15NO3
InChi Key: GDPSCBPOCONUDM-UHFFFAOYNA-N
SMILES: O=CC1CCN(C1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results